CaCO₃ in Salpetersaure 0.1 % ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF21531.0000
Supplier: Bernd Kraft
CaCO₃ in Salpetersaure 0.1 % ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF21531.0000
Supplier: Bernd Kraft
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur. 1091201
Supplier: VWR Chemicals
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
ISE standards
Supplier: Hach
Ammonium dihydrogen phosphate, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach