Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Sodium arsenite solution 2 g/l for determination of free chlorine and total chlorine according to EN ISO 7393:2000, DEV G4 Content: 0,5 l 1 * 500 mL
KRAF20711.2000
:
Bernd Kraft
Sodium arsenite solution 2 g/l for determination of free chlorine and total chlorine according to EN ISO 7393:2000, DEV G4 Content: 0,5 l 1 * 500 mL
KRAF20711.2000
:
Bernd Kraft