Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 5%, extra pure

Ammonia solution 5%, extra pure

Plastic bottle, 2.500 mL
Artikelnummer KRAF12388.4000
Irritant
Hersteller:   Bernd Kraft
Ammonia solution 5%, extra pure
Artikelnummer KRAF12388.4000
Hersteller:  Bernd Kraft
CAS-Nummer:  1336-21-6
Ammonia solution 5%, extra pure

Plastic bottle, 2.500 mL

CAS-Nummer:  1336-21-6
Preis
65,20 €
Log in for availability
1 * 2.500 mL
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-647-6
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2.500 mL

Specifications