Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 5% analytical reagent

Ammonia solution 5% analytical reagent

Plastic bottle, 10 L
Catalog # KRAF12389.6020
Irritant
Supplier:   Bernd Kraft
Ammonia solution 5% analytical reagent
Catalog # KRAF12389.6020
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 5% analytical reagent

Plastic bottle, 10 L

CAS Number:  1336-21-6
Price
185,00 €
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-647-6
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 L

Specifications