Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hydroxide 4% in aqueous solution, Technical Grade

Sodium hydroxide 4% in aqueous solution, Technical Grade

Plastic bottle, 20 L
Catalog # KRAF11431.7020
Corrosive
Supplier:   Bernd Kraft
Sodium hydroxide 4% in aqueous solution, Technical Grade
Catalog # KRAF11431.7020
Supplier:  Bernd Kraft
CAS Number:  1310-73-2
Sodium hydroxide 4% in aqueous solution, Technical Grade

Plastic bottle, 20 L

CAS Number:  1310-73-2
Price
145,00 €
Log in for availability
1 * 20 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    20 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.