Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hydroxide 4% in aqueous solution, Technical Grade

Sodium hydroxide 4% in aqueous solution, Technical Grade

Plastic bottle, 5 L
Catalog # KRAF11431.5020
Corrosive
Supplier:   Bernd Kraft
Sodium hydroxide 4% in aqueous solution, Technical Grade
Catalog # KRAF11431.5020
Supplier:  Bernd Kraft
CAS Number:  1310-73-2
Sodium hydroxide 4% in aqueous solution, Technical Grade

Plastic bottle, 5 L

CAS Number:  1310-73-2
Price
61,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications