Orders LinkContactUsLinkComponent
Ammonia solution 20%, Technical Grade
Plastic bottle, 25 L
  KRAF20073.7910
Ammonia solution 20%, Technical Grade
  KRAF20073.7910
 :  1336-21-6
Ammonia solution 20%, Technical Grade
Plastic bottle, 25 L
 :  1336-21-6
185,00 €
1 * 25 L
 :  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  •  :1336-21-6
  •  :NH₃
  •  :MFCD00066650
  •  :35.05 g/mol
  •  :0.92 g/cm³ (20 °C)
  •  :Ambient
  •  :215-647-6
  •  :2672
  •  :8,III
  • Pack type:
    Plastic bottle
  • Pk:
    25 L