Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 20%, Technical Grade

Ammonia solution 20%, Technical Grade

Plastic bottle, 5 L
Catalog # KRAF20073.5010
Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 20%, Technical Grade
Catalog # KRAF20073.5010
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 20%, Technical Grade

Plastic bottle, 5 L

CAS Number:  1336-21-6
Price
74,10 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.