Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ammonia solution 25%, Technical Grade

Plastic bottle, 10 L
Catalog # KRAF06110.6010
Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 25%, Technical Grade
Catalog # KRAF06110.6010
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 25%, Technical Grade

Plastic bottle, 10 L

CAS Number:  1336-21-6
Price
61,30 €
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 L

Specifications