Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 32%, Technical Grade

Ammonia solution 32%, Technical Grade

Plastic bottle, 5 L
Catalog # KRAF20070.5010
Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 32%, Technical Grade
Catalog # KRAF20070.5010
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 32%, Technical Grade

Plastic bottle, 5 L

CAS Number:  1336-21-6
Price
106,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications