Order Entry
Germany
Orders LinkContactUsLinkComponent
Diethyl ether stabilised, extra pure

Diethyl ether stabilised, extra pure

Glass bottle, 500 mL
Catalog # KRAF07106.2700
Flammable
Irritant
Supplier:   Bernd Kraft
Diethyl ether stabilised, extra pure
Catalog # KRAF07106.2700
Supplier:  Bernd Kraft
CAS Number:  60-29-7
Diethyl ether stabilised, extra pure
Diethyl ether stabilised, extra pure

Glass bottle, 500 mL

CAS Number:  60-29-7
Price
33,70 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 7 ppm
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications