Diethyl ether stabilised analytical reagent
25 L
Catalog # KRAF21161.8900
Supplier: Bernd Kraft
Diethyl ether stabilised analytical reagent
Catalog # KRAF21161.8900
Supplier: Bernd Kraft
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0.7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- UN:1155
- ADR:3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
- Storage Temperature:Ambient
- EINECS:200-467-2
- REACH:01-2119535785-29
Specifications
Specifications
- Pk:25 L



