Order Entry
Germany
Orders LinkContactUsLinkComponent
Diethyl ether stabilised analytical reagent

Diethyl ether stabilised analytical reagent

Glass bottle, 1 L
Catalog # KRAF21161.3700
Flammable
Irritant
Supplier:   Bernd Kraft
Diethyl ether stabilised analytical reagent
Catalog # KRAF21161.3700
Supplier:  Bernd Kraft
CAS Number:  60-29-7
Diethyl ether stabilised analytical reagent
Diethyl ether stabilised analytical reagent

Glass bottle, 1 L

CAS Number:  60-29-7
Price
141,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.