Cyclohexane, Technical Grade
Plastic bottle, 10 L
KRAF20149.6700
: Hexahydrobenzene, Hexamethylene
- :110-82-7
- :C₆H₁₂
- :MFCD00003814
- :84.16 g/mol
- :13,02752
- :81 °C (1013 hPa)
- :7 °C
- :0.7785 g/cm³ (20 °C)
- :–18 °C
- :Ambient
- :203-806-2
- :1145
- :3,II
- Pack type:Plastic bottle
- Pk:10 L