Order Entry
Germany
Orders LinkContactUsLinkComponent
Cyclohexane, extra pure

Cyclohexane, extra pure

Plastic bottle, 10 L
Catalog # KRAF07161.6700
Flammable
Health
Irritant
Environment
Supplier:   Bernd Kraft
Cyclohexane, extra pure
Catalog # KRAF07161.6700
Supplier:  Bernd Kraft
CAS Number:  110-82-7
Cyclohexane, extra pure
Cyclohexane, extra pure

Plastic bottle, 10 L

CAS Number:  110-82-7
Price
180,00 €
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.