Order Entry
Germany
Orders LinkContactUsLinkComponent
Cyclohexane, extra pure

Cyclohexane, extra pure

5 L
Catalog # KRAF07161.5500
Flammable
Health
Irritant
Environment
Supplier:   Bernd Kraft
Cyclohexane, extra pure
Catalog # KRAF07161.5500
Supplier:  Bernd Kraft
CAS Number:  110-82-7
Cyclohexane, extra pure
Cyclohexane, extra pure

5 L

CAS Number:  110-82-7
Price
110,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications

Specifications

  • Pk:
    5 L

Specifications