Chloroform stabilised, Technical Grade
Glass bottle, 2.500 mL
KRAF20100.4700
: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
- :67-66-3
- :CHCl₃
- :MFCD00000826
- :119.38 g/mol
- :13,02160
- :61.7 °C (1013 hPa)
- :–63 °C
- :1.4832 g/cm³ (20 °C)
- :200-663-8
- :6.1,III
- :Stabilised with ethanol
- :Ambient
- :1888
- :05-2114614906-45
- Pack type:Glass bottle
- Pk:2.500 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



