Order Entry
Germany
Orders LinkContactUsLinkComponent
Chloroform stabilised, Technical Grade

Chloroform stabilised, Technical Grade

Glass bottle, 1 L
Catalog # KRAF20100.3700
Toxic
Health
Supplier:   Bernd Kraft
Chloroform stabilised, Technical Grade
Catalog # KRAF20100.3700
Supplier:  Bernd Kraft
CAS Number:  67-66-3
Chloroform stabilised, Technical Grade
Chloroform stabilised, Technical Grade

Glass bottle, 1 L

CAS Number:  67-66-3
Price
37,50 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • EINECS:200-663-8
  • ADR:6.1,III
  •  :Stabilised with ethanol
  • Storage Temperature:Ambient
  • UN:1888
  • REACH:05-2114614906-45
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications