Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium nitrite, extra pure

Sodium nitrite, extra pure

Plastic bottle for solids, 500 g
Catalog # KRAF06201.2600
Oxidizing
Toxic
Environment
Supplier:   Bernd Kraft
Sodium nitrite, extra pure
Catalog # KRAF06201.2600
Supplier:  Bernd Kraft
CAS Number:  7632-00-0
Sodium nitrite, extra pure

Plastic bottle for solids, 500 g

CAS Number:  7632-00-0
Price
34,30 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Diazotizing salts
  • CAS Number:7632-00-0
  • Formula:NaNO₂
  • MDL Number:MFCD00011118
  • Molecular Weight:69 g/mol
  • Merck Index:13,08721
  • Boiling Pt:320 °C (1013 hPa)
  • Melting Pt:284 °C
  • Density:2.168 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-555-9
  • UN:1500
  • ADR:5.1,III
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.