0.1 mol ICl/l - 0.2 N Losung in Essigsaure zur Bestimmung der Iodzahl gemaß DIN 53241-1:1995 1 * 500 mL
Catalog # KRAF05171.2700
Supplier: Bernd Kraft
0.1 mol ICl/l - 0.2 N Losung in Essigsaure zur Bestimmung der Iodzahl gemaß DIN 53241-1:1995 1 * 500 mL
Catalog # KRAF05171.2700
Supplier: Bernd Kraft
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sterican® Single-Use Needles for Special Applications
Supplier: B. Braun