Acetone, Technical Grade
Plastic bottle, 5 L
Supplier: Bernd Kraft
Acetone, Technical Grade
Catalog # KRAF13455.5300
Supplier: Bernd Kraft
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56.2 °C (1013 hPa)
- Melting Pt:–95.4 °C
- Density:0.792 g/cm³ (20 °C)
- Flash Pt:<–20 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:5 L
- Cat. No.:KRAF13455.5300
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



