Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solution, colour coded, ready to use

10,0, Borate, Blue, 20 °C, Plastic bottle, 10 L
Catalog # KRAF10581.6000
Supplier:   Bernd Kraft
Buffer solution, colour coded, ready to use
Catalog # KRAF10581.6000
Supplier:  Bernd Kraft
CAS Number:  
Buffer solution, colour coded, ready to use
Buffer solution, colour coded, ready to use

10,0, Borate, Blue, 20 °C, Plastic bottle, 10 L

CAS Number:  
Price
189,00 €
Log in for availability
1 * 10 L
Product Details & Documents
  • NIST traceable
  • Colour coded with choice of colours for pH values 4,00 and 7,00
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    10 L
  • Reference temperature:
    20 °C
  • Colour:
    Blue
  • pH value:
    10,0

Specifications