Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solution, colour coded, ready to use

Buffer solution, colour coded, ready to use

10,0, Borate, Blue, 20 °C, Plastic bottle, 1 L
Catalog # KRAF10581.3000
Supplier:   Bernd Kraft
Buffer solution, colour coded, ready to use
Catalog # KRAF10581.3000
Supplier:  Bernd Kraft
CAS Number:  
Buffer solution, colour coded, ready to use
Buffer solution, colour coded, ready to use

10,0, Borate, Blue, 20 °C, Plastic bottle, 1 L

CAS Number:  
Price
40,80 €
Log in for availability
1 * 1 L
Product Details & Documents
  • NIST traceable
  • Colour coded with choice of colours for pH values 4,00 and 7,00
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • Colour:
    Blue
  • pH value:
    10,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.