Buffer solution, colour coded, ready to use
7,0, Phosphate, Green, 20 °C, Plastic bottle, 10 L
Catalog # KRAF03786.6000
Supplier: Bernd Kraft
Buffer solution, colour coded, ready to use
Catalog # KRAF03786.6000
Supplier: Bernd Kraft
CAS Number:
Jump to:
Product Details & Documents
- NIST traceable
- Colour coded with choice of colours for pH values 4,00 and 7,00
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:10 L
- Reference temperature:20 °C
- Colour:Green
- pH value:7,0



