Buffer solution, colour coded, ready to use
4,0, Citrate, Green, 20 °C, Plastic bottle, 500 mL
Catalog # KRAF03787.2000
Supplier: Bernd Kraft
Buffer solution, colour coded, ready to use
Catalog # KRAF03787.2000
Supplier: Bernd Kraft
CAS Number:
Jump to:
Product Details & Documents
- NIST traceable
- Colour coded with choice of colours for pH values 4,00 and 7,00
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:500 mL
- Reference temperature:20 °C
- Colour:Green
- pH value:4,0
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



