Buffer solution, colour coded, ready to use
4,0, Citrat, Rot, 20 °C, Kunststoffflasche, 5 L
Supplier: Bernd Kraft
Buffer solution, colour coded, ready to use
Catalog # KRAF03283.5400
Supplier: Bernd Kraft
CAS Number:
Jump to:
Product Details & Documents
- NIST traceable
- Colour coded with choice of colours for pH values 4,00 and 7,00
Specifications
Specifications
- Packung:Kunststoffflasche
- pH-Puffertyp:Citrat
- VE:5 L
- Referenztemperatur:20 °C
- Farbe:Rot
- pH-Wert:4,0



