Order Entry
Germany
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid analytical reagent

L(+)-Ascorbic acid analytical reagent

Plastic bottle for solids, 1 kg
Catalog # KRAF19297.3600
Supplier:   Bernd Kraft
L(+)-Ascorbic acid analytical reagent
Catalog # KRAF19297.3600
Supplier:  Bernd Kraft
CAS Number:  50-81-7
L(+)-Ascorbic acid analytical reagent
L(+)-Ascorbic acid analytical reagent

Plastic bottle for solids, 1 kg

CAS Number:  50-81-7
Price
220,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.