Buffer solution, colour coded, ready to use
2,0, Citrate, Yellow, 20 °C, Plastic bottle, 10 L
KRAF10926.6400
- NIST traceable
- Colour coded with choice of colours for pH values 4,00 and 7,00
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:10 L
- Reference temperature:20 °C
- Colour:Yellow
- pH value:2,0



