Buffer solution, colour coded, ready to use
2,0, Citrate, Yellow, 20 °C, Plastic bottle, 0,5 L
KRAF10926.2000
- NIST traceable
- Colour coded with choice of colours for pH values 4,00 and 7,00
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:0,5 L
- Reference temperature:20 °C
- Colour:Yellow
- pH value:2,0
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



