Buffer solutions, ready to use
13,0, Glycine, Colourless, 20 °C, Plastic bottle, 5 L
Catalog # KRAF03096.5400
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
- NIST traceable
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Glycine
- Pk:5 L
- Reference temperature:20 °C
- Colour:Colourless
- pH value:13,0



