Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solutions, ready to use

Buffer solutions, ready to use

9,0, Borate, Colourless, 20 °C, Plastic bottle, 5 L
Artikelnummer KRAF03088.5400
Hersteller:   Bernd Kraft
Buffer solutions, ready to use
Artikelnummer KRAF03088.5400
Hersteller:  Bernd Kraft
CAS-Nummer:  
Buffer solutions, ready to use
Buffer solutions, ready to use

9,0, Borate, Colourless, 20 °C, Plastic bottle, 5 L

CAS-Nummer:  
Preis
139,00 €
Log in for availability
1 * 5 L
Product Details & Documents
  • NIST traceable

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    5 L
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    9,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.