Buffer solutions, ready to use
8,0, Phosphate, Colourless, 20 °C, Plastic bottle, 10 L
Catalog # KRAF03087.6000
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
- NIST traceable
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:10 L
- Reference temperature:20 °C
- Colour:Colourless
- pH value:8,0
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



