Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

3,0, Citrate, Colourless, 20 °C, Plastic bottle, 5 L
Catalog # KRAF03082.5400
Supplier:   Bernd Kraft
Buffer solutions, ready to use
Catalog # KRAF03082.5400
Supplier:  Bernd Kraft
CAS Number:  
Buffer solutions, ready to use
Buffer solutions, ready to use

3,0, Citrate, Colourless, 20 °C, Plastic bottle, 5 L

CAS Number:  
Price
139,00 €
Log in for availability
1 * 5 L
Product Details & Documents
  • NIST traceable

 

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Citrate
  • Pk:
    5 L
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    3,0

Specifications