Order Entry
Germany
Orders LinkContactUsLinkComponent
Formaldehyde 37% in aqueous solution stabilised, extra pure

Formaldehyde 37% in aqueous solution stabilised, extra pure

Plastic bottle, 2.500 mL
Catalog # KRAF04038.4000
Toxic
Corrosive
Health
Supplier:   Bernd Kraft
Formaldehyde 37% in aqueous solution stabilised, extra pure
Catalog # KRAF04038.4000
Supplier:  Bernd Kraft
CAS Number:  50-00-0
Formaldehyde 37% in aqueous solution stabilised, extra pure

Plastic bottle, 2.500 mL

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 2.500 mL
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • ADR:8,III
  •  :Stabilised with methanol
  • EINECS:200-001-8
  • UN:2209
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.