ContactUsLinkComponent
Filter, CHROMAFIL disposable RC-20/15mS membrane, material:regenerated cellulose pore diameter:0.2µm, membrane diameter:15mm colour code above:yellow / below:blue pack of 800. 1 * 800 items
Catalog # MANA729036.800
Lieferant:  MACHEREY-NAGEL
undefined
Filter, CHROMAFIL disposable RC-20/15mS membrane, material:regenerated cellulose pore diameter:0.2µm, membrane diameter:15mm colour code above:yellow / below:blue pack of 800. 1 * 800 items
Catalog # MANA729036.800
Lieferant:  MACHEREY-NAGEL
Frequently Bought Together