Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
REFRACTOMER, POCKET DIGITAL, BRAKE-CHEK WITH MULTI-SCALE MEASUREMENT FEATURE. MEASURE BOILING POINT OF DOT3, DOT3 HIGH TEMP, DOT4, DOT4 PLUS BRAKE FLUIDS - FAHRENHEIT SCALE 1 * 1 items
REIC13940016
:
REICHERT
REFRACTOMER, POCKET DIGITAL, BRAKE-CHEK WITH MULTI-SCALE MEASUREMENT FEATURE. MEASURE BOILING POINT OF DOT3, DOT3 HIGH TEMP, DOT4, DOT4 PLUS BRAKE FLUIDS - FAHRENHEIT SCALE 1 * 1 items
REIC13940016
:
REICHERT