Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Bottles, narrow neck, with thread

300 ml
Catalog # 215-3230
Supplier:   GRAVIS
Bottles, narrow neck, with thread
Catalog # 215-3230
Supplier:  GRAVIS
Bottles, narrow neck, with thread
Bottles, narrow neck, with thread

300 ml

Price
110,00 €
Log in for availability
1 * 48 ST
Product Details & Documents

Glass, clear, amber or yellow.

  • Neck Ø: 28 mm
  • Good chemical and temperature resistance
  • Amber coloured to protect light sensitive substances

Choice of various screw caps.

 : Supplied without screw caps - please order separately.

 : Packaged in impermeable plastic packs.

Specifications
  • Capacity:
    300 ml
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.