Order Entry
Germany
Orders LinkContactUsLinkComponent
Heating tapes, HST

Heating tapes, HST

Heating tape HST with 10 m length, 480 W output
Catalog # HORS025207
Supplier:   HORST
Heating tapes, HST
Catalog # HORS025207
Supplier:  HORST
Heating tapes, HST

Heating tape HST with 10 m length, 480 W output

Price
368,78 €
Log in for availability
1 * 1 ST
Product Details & Documents

These slim heating tapes consist of two PTFE insulated heating conductors with earthed protection braid which have a common coating made of metal cloth.

Specifications
  • Description:
    Heating tape HST with 10 m length, 480 W output
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.