Ammonia solution 5%, extra pure
1
Catalog # KRAF12388.3000
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- CAS Number:1336-21-6
- Formula:NH₃
- MDL Number:MFCD00066650
- Molecular Weight:35.05 g/mol
- Storage Temperature:Ambient
- EINECS:215-647-6
Specifications
Specifications
- Pk:1



