Cell counter vials
Cell counter vial 2 without lid, for cell counter systems with cylindrical containers, e.g. Al® and Linson®, 20,00 ml, PS, Clear, No, 1000 vials/bag
Catalog # RATI5515051
Supplier: RATIOLAB
Jump to:
Product Details & Documents
Cell counter vials made of clear PS.
- High-precision design for use with robotic analytical systems
Specifications
- Cap included:No
- Packed:1000 vials/bag
- Capacity:20,00 ml
- Colour:Clear
- Description:Cell counter vial 2 without lid, for cell counter systems with cylindrical containers, e.g. Al® and Linson®
- Material:PS
- Seq:04