Orders LinkContactUsLinkComponent
Stoppers, for test tubes
Stoppers, for test tubes
Blue, 11 - 12 mm
Catalog # RATI351490B
Supplier:   RATIOLAB
Stoppers, for test tubes
Catalog # RATI351490B
Supplier:  RATIOLAB
Stoppers, for test tubes
Stoppers, for test tubes
Blue, 11 - 12 mm
Price
17,20 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
PE.
  • Very tight seal due to two sealing rings
  • Available in natural and four more colors
Specifications
  • Ø bottom:
    11 - 12 mm
  • Colour:
    Blue
  • Seq:
    02