Order Entry
Germany
Orders LinkContactUsLinkComponent
Multi purpose bottles

Multi purpose bottles

2500 ml, Multipurpose collecting bottles with label, Blue, 80 mm
Catalog # RATI5922250
Supplier:   RATIOLAB
Multi purpose bottles
Catalog # RATI5922250
Supplier:  RATIOLAB
Multi purpose bottles
Multi purpose bottles

2500 ml, Multipurpose collecting bottles with label, Blue, 80 mm

Price
171,98 €
Log in for availability
1 * 60 ST
Product Details & Documents

Polyethylene, blue translucent.

  • Incl. lid with nonwoven fabric liner
  • With graduations
  • Light protection
  • Wide neck diameter of 80 mm
Specifications
  • Capacity:
    2500 ml
  • Colour:
    Blue
  • Description:
    Multipurpose collecting bottles with label
  • Seq:
    01
  • Neck-Ø:
    80 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.