Stoppers, for test tubes
Natural, 11 - 12 mm
Catalog # RATI3514900
Supplier: RATIOLAB
Jump to:
Product Details & Documents
PE.
- Very tight seal due to two sealing rings
- Available in natural and four more colors
Specifications
- Ø bottom:11 - 12 mm
- Colour:Natural
- Seq:01