Orders LinkContactUsLinkComponent
Drum stands, FB
Drum stands, FB
Drum stands, FB 3, fire-galvanised with roller support, capacity 3×60 l drum, 1230×650×415 mm
Catalog # LACOP21-1803-A
Drum stands, FB
Catalog # LACOP21-1803-A
Drum stands, FB
Drum stands, FB 3, fire-galvanised with roller support, capacity 3×60 l drum, 1230×650×415 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Drum stands with fixed drum supports for 60 to 200 L drums, completely galvanised. Tilted support onwards for easy drum emptying.
These drum stands can be combined with compact sump pallets, water protection pallets, hazardous substance stations, floor safety elements and drum carts.
Specifications
  • Description:
    Drum stands, FB 3, fire-galvanised with roller support, capacity 3×60 l drum
  • Seq:
    06
  • W×D×H:
    1230×650×415 mm