Drum stands, FB
Drum stands, FB 2, fire-galvanised with roller support, capacity 2×200 l drum, 1230×650×415 mm
Catalog # LACOP21-1802-A
Supplier: LACONT UMWELTTECHNIK
Jump to:
Product Details & Documents
Drum stands with fixed drum supports for 60 to 200 L drums, completely galvanised. Tilted support onwards for easy drum emptying.
These drum stands can be combined with compact sump pallets, water protection pallets, hazardous substance stations, floor safety elements and drum carts.
Specifications
- Description:Drum stands, FB 2, fire-galvanised with roller support, capacity 2×200 l drum
- Seq:04
- W×D×H:1230×650×415 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



