Accessories for fluid and solid samplers
Cleaning brush, PVC/stainless steel, Ø 25 mm, 400 mm
300-0109
: BURKLE
- Description:Cleaning brush, PVC/stainless steel, Ø 25 mm, 400 mm
- Seq:1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



