Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Stoppers, chloroprene rubber

65,0 mm, 94,0 mm, 107,0 mm
Catalog # DENE1010494
Supplier:   DEUTSCH NEUMANN
Stoppers, chloroprene rubber
Catalog # DENE1010494
Supplier:  DEUTSCH NEUMANN
Stoppers, chloroprene rubber
Stoppers, chloroprene rubber

65,0 mm, 94,0 mm, 107,0 mm

Price
231,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
  • Durometer 40 Shore A

 : Acc. DIN 12 871

Specifications
  • Ø bottom:
    94,0 mm
  • Ø top:
    107,0 mm
  • Height:
    65,0 mm