Orders LinkContactUsLinkComponent
Hypodermic needles
Hypodermic needles
Hypodermic, sharp, 18, 150 mm
Catalog # MURRM618/08
Supplier:   MURRAY S
Hypodermic needles
Catalog # MURRM618/08
Supplier:  MURRAY S
Hypodermic needles
Hypodermic needles
Hypodermic, sharp, 18, 150 mm
Price
54,70 €
Log in for availability
1 * 10 ST
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specifications
  • Gauge:
    18
  • Description:
    Hypodermic, sharp
  • Length:
    150 mm
  • Seq:
    16