Hypodermic needles
Hypodermic, sharp, 18, 50 mm
Catalog # MURRM618/03
Supplier: MURRAY S
Jump to:
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specifications
- Gauge:18
- Description:Hypodermic, sharp
- Length:50 mm
- Seq:13