Orders LinkContactUsLinkComponent
Hypodermic needles
Hypodermic, sharp, 16, 100 mm
Catalog # MURRM616/06
Supplier:   MURRAY S
Hypodermic needles
Catalog # MURRM616/06
Supplier:  MURRAY S
Hypodermic needles
Hypodermic needles
Hypodermic, sharp, 16, 100 mm
Price
54,10 €
Log in for availability
1 * 10 ST
Product Details & Documents
Non sterile needles with sharp ends and luer lock fitting.
Specifications
  • Gauge:
    16
  • Description:
    Hypodermic, sharp
  • Length:
    100 mm
  • Seq:
    11